1-Iodododecane, 98%, stabilized
1-Iodododecane, 98%, stabilized
1-Iodododecane, 98%, stabilized
Thermo Scientific Chemicals

1-Iodododecane, 98%, stabilized

CAS: 4292-19-7 | C12H25I | 296.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
View all
Glass bottle
Catalog number 250091000
Price (EUR)
120,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
120,00
Each
Chemical Identifiers
CAS4292-19-7
IUPAC Name1-iodododecane
Molecular FormulaC12H25I
InChI KeyGCDPERPXPREHJF-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
StabilizerCopper
Appearance (Form)Liquid
Refractive index1.4825 to 1.4865 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only