Arecoline hydrobromide, 98+%
Arecoline hydrobromide, 98+%
Arecoline hydrobromide, 98+%
Thermo Scientific Chemicals

Arecoline hydrobromide, 98+%

CAS: 300-08-3 | C8H14BrNO2 | 236.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 250130250
Price (EUR)
125,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
125,00
Each
Chemical Identifiers
CAS300-08-3
IUPAC Namehydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
Molecular FormulaC8H14BrNO2
InChI KeyAXOJRQLKMVSHHZ-UHFFFAOYSA-N
SMILES[H+].[Br-].COC(=O)C1=CCCN(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric>=98.0 %
Appearance (Color)White to light yellow
Melting point168°C to 174°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only