Etil 2-metilbutirato, 99 %, Thermo Scientific Chemicals
Etil 2-metilbutirato, 99 %, Thermo Scientific Chemicals
Etil 2-metilbutirato, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Etil 2-metilbutirato, 99 %, Thermo Scientific Chemicals

CAS: 7452-79-1 | C7H14O2 | 130.19 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 250261000
Precio (EUR)
51,10
Each
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
51,10
Each
Identificadores químicos
CAS7452-79-1
IUPAC Nameethyl 2-methylbutanoate
Molecular FormulaC7H14O2
InChI KeyHCRBXQFHJMCTLF-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)CC
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3950 to 1.3990 (20°C, 589 nm)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only