L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
Thermo Scientific Chemicals

L-Lactic acid, sodium salt, 60 wt% solution in water

CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2503000101 LGlass bottle
Catalog number 250300010
Price (TWD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS867-56-1
IUPAC Namesodium 2-hydroxypropanoate
Molecular FormulaC3H5NaO3
InChI KeyNGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES[Na+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear viscous liquid
Refractive index1.4220 to 1.4250 (20°C, 589 nm)
Density(20°C) 1.32 to 1.34 g/ml
Titration with HCl59 to 61 %
Sodium (Na)12.1 to 12.6 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only