Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets BP, EP and USP requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
500 mL
2.5 kg
2.5 L
12 kg
40 lb.
175 lb.
450 lb.
Packaging:
View all
Polyethylene drum, open head
Catalog number 2504-08
Price (MXN)
-
Quantity:
175 lb.
Packaging:
Polyethylene drum, open head
This product is GMP Manufactured and meets BP, EP and USP requirements.
This product is GMP Manufactured and meets BP, EP and USP requirements.
Specifications
CAS10034-99-8
Chemical Name or MaterialMagnesium sulfate heptahydrate
ColorColorless
Melting Point1124°C
Molecular FormulaMgSO4·7H2O
PackagingPolyethylene drum, open head
SynonymEpsom salts
FormCrystalline Powder
GradeMulti-Compendial/USP
Molecular Weight (g/mol)246.48
Percent Purity99 to 100.5%
Quantity175 lb.
pH5 to 9.2 (25°C, 5% aqueous solution)
Unit SizeEach