Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets BP, EP and USP requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
500 mL
2.5 kg
2.5 L
12 kg
40 lb.
175 lb.
450 lb.
Packaging:
View all
Clear glass bottle
Catalog number 2504-44
Price (CLP)
-
Request A Quote
Quantity:
2.5 L
Packaging:
Clear glass bottle
This product is GMP Manufactured and meets BP, EP and USP requirements.
This product is GMP Manufactured and meets BP, EP and USP requirements.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS10034-99-8
Chemical Name or MaterialMagnesium sulfate heptahydrate
ColorColorless
Melting Point1124°C
Molecular FormulaMgSO4·7H2O
PackagingClear glass bottle
SynonymEpsom salts
FormCrystalline Powder
Molecular Weight (g/mol)246.48
Percent Purity99 to 100.5%
Quantity2.5 L
pH5 to 9.2 (25°C, 5% aqueous solution)
Unit SizeEach