Nalgene™ PPCO Dilution Bottles with Closure, 200mL - FAQs
View additional product information for Nalgene™ PPCO Dilution Bottles with Closure - FAQs (2505-0280PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.