Product Image
Thermo Scientific Chemicals

L-Dehydroascorbic acid, 96%

CAS: 490-83-5 | C6H6O6 | 174.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 250932500
Price (USD)
58.90
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
58.90
Each
Chemical Identifiers
CAS490-83-5
IUPAC Name5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
Molecular FormulaC6H6O6
InChI KeySBJKKFFYIZUCET-UHFFFAOYNA-N
SMILESOCC(O)C1OC(=O)C(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to light brown
Infrared spectrumConforms
Titration with NaOH95 to 105 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only