25103-032130 Full specifications
Each 3 μm 2.1 mm 30 mm Request A Quote - Column Format Analytical Column Diameter (Metric) 2.1 mm For Use With HPLC Length (Metric) 30 mm Max. Pressure 5800 psi (400 bar) Particle Size 3 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25105-154630 Full specifications
Each 5 μm 4.6 mm 150 mm Column Format Analytical Column Diameter (Metric) 4.6 mm Length (Metric) 150 mm Max. Pressure 5800 psi (400 bar) Particle Size 5 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25105-253030 Full specifications
Each 5 μm 3 mm 250 mm Contact Us › - Column Format Analytical Column Diameter (Metric) 3 mm Length (Metric) 250 mm Max. Pressure 5800 psi (400 bar) Particle Size 5 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25105-254030 Full specifications
Each 5 μm 4 mm 250 mm Request A Quote - Column Format Analytical Column Diameter (Metric) 4 mm Length (Metric) 250 mm Max. Pressure 5800 psi (400 bar) Particle Size 5 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25105-254630 Full specifications
Each 5 μm 4.6 mm 250 mm Column Format Analytical Column Diameter (Metric) 4.6 mm Length (Metric) 250 mm Max. Pressure 5800 psi (400 bar) Particle Size 5 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25102-202130 Full specifications
Each 1.9 μm 2.1 mm 200 mm Column Format Analytical Column Diameter (Metric) 2.1 mm For Use With HPLC Length (Metric) 200 mm Max. Pressure 18130 psi (1250 bar) Particle Size 1.9 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25102-152130 Full specifications
Each 1.9 μm 2.1 mm 150 mm Request A Quote - Column Format Analytical Column Diameter (Metric) 2.1 mm For Use With HPLC Length (Metric) 150 mm Max. Pressure 18130 psi (1250 bar) Particle Size 1.9 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25102-102130 Full specifications
Each 1.9 μm 2.1 mm 100 mm Column Format Analytical Column Diameter (Metric) 2.1 mm For Use With HPLC Length (Metric) 100 mm Max. Pressure 18130 psi (1250 bar) Particle Size 1.9 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25102-052130 Full specifications
Each 1.9 μm 2.1 mm 50 mm Request A Quote - Column Format Analytical Column Diameter (Metric) 2.1 mm For Use With HPLC Length (Metric) 50 mm Max. Pressure 18130 psi (1250 bar) Particle Size 1.9 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each
25103-154630 Full specifications
Each 3 μm 4.6 mm 150 mm Column Format Analytical Column Diameter (Metric) 4.6 mm For Use With HPLC Length (Metric) 150 mm Max. Pressure 5800 psi (400 bar) Particle Size 3 μm pH 2 to 8 Pore Size 175 Å Surface Area 220 m²/g Temperature 60°C USP Type L3 Stationary Phase Silica Product Line Hypersil GOLD Packing Material Spherical, Fully Porous, Ultrapure Silica Column Type Normal Phase Endcapped No Phase Normal Phase Type Analytical Column Unit Size Each