Search
View additional product information for Nunc™ Amplification Workstation - FAQs (251357)
2 product FAQs found
Yes, our Nunc Amplification Workstation is autoclavable. The acceptable temperature range is from -20 degrees C to 130 degrees C.
Our Nunc Amplification Workstation is made of polyphenylenine oxide. This material is autoclavable and is resistant to solvents and weak acids.