This product will be unavailable for purchase after April 1, 2016, or when stock is depleted, whichever comes first. We recommend Cat. No. XP00082BOX as a possible alternative, depending on the gel tank being used. Learn about our Novex WedgeWell Tris-Glycine gels. Please contact us with any questions.

Precise™ 8% Tris-Glycine Gels, 6.8 cm x 8 cm x 1 mm, 12-well

Catalog number: 25265

Thermo Scientific™  Related applications: Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Precise 8% Tris-Glycine Gels, 6.8 cm x 8 cm x 1 mm, 12-well, are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Features Precise Tris-Glycine Gels:

High-performance—Tris-glycine running buffer produces excellent separation and high-resolution protein bands
Long shelf-life—up to 12-month stability ensures consistent performance for the duration of each package of gels
Fast—45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains
Compatible— these 8 cm x 10 cm x 5 mm cassettes fit Bio-Rad™ Mini-PROTEAN™ and Hoefer™ Mighty Small™ II gel tanks, as well as other similar tanks
Easy to load—sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Precise Tris-Glycine Gels are cast in durable 8 cm x 10 cm x 5 mm plastic cassettes to fit in Mini-PROTEAN™ XCell gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools.

Specifications:
Product Details:
Precise Tris-Glycine Gels were developed to work with the Laemmli Tris-glycine running buffer. Unlike other Tris-glycine SDS-PAGE gels, Precise Tris-Glycine Gels are cast in a neutral buffer system, which allows them to have a long shelf-life. Furthermore, Precise Tris-Glycine Gels can withstand the chemical changes caused by the Laemmli running-buffer that alter gel conductivity and increase run-times in other Tris-glycine gels. As a result, Precise Tris-Glycine Gels allow users to achieve excellent protein resolution in a shorter time.

Related Products
Precise™ 8% Tris-Glycine Gels, 6.8 cm x 8 cm x 1 mm, 10-well
Precise™ 8% Tris-Glycine Gels, 6.8 cm x 8 cm x 1 mm, 15-well
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gel Percentage: 8 %
Gel Size: Mini (6.8 cm x 8 cm)
Gel Type: Tris-Glycine Gel
Mode of Separation: Molecular Weight
Product Line: Precise™
Product Size: 10 gels (1 box)
Quantity: 10 gels
Separation Range: 40 kDa to 200 kDa
Thickness: 1.0 mm
Well Format: 1D Well
Wells: 12 wells

Contents & storage

One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 12 months.

Documents

Manuals & protocols