Lead(II) perchlorate trihydrate, 99%
Lead(II) perchlorate trihydrate, 99%
Lead(II) perchlorate trihydrate, 99%
Thermo Scientific Chemicals

Lead(II) perchlorate trihydrate, 99%

CAS: 13453-62-8 | Cl2H6O11Pb | 460.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 254590250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13453-62-8
IUPAC Nameλ²-lead(2+) trihydrate diperchlorate
Molecular FormulaCl2H6O11Pb
InChI KeyKKGGEAQRICYXNM-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Adhering crystals
Titration Complexometric>=98.5 %
pH3 to 5 (5 % solution)
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only