Hypersil GOLD™ PREP C4 XL HPLC Column Repacking Service
Thermo Scientific™

Hypersil GOLD™ PREP C4 XL HPLC Column Repacking Service

Ensure optimal chromatography performance and cost savings with Hypersil GOLD PREP C4 XL HPLC column repacking services.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
25505-109270XL-R25 mm100 mm
25505-259270XL-R25 mm250 mm
Catalog number 25505-109270XL-R
Price (USD)
-
Diameter (Metric):
25 mm
Length (Metric):
100 mm
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil GOLD™ PREP C4 XL HPLC column repacking service. Choose ExtendedLife (XL) preparative column hardware during your initial column purchase to be eligible for repacking services. All XL preparative columns are equipped with Dynamic Axial Compression technology to increase column lifetime. Return your XL column to Thermo Fisher to have it refreshed with short alkyl chain length, low hydrophobicity C4 media without paying for new hardware.

XL Column Repacking Service

Choose XtendedLife Hardware at Initial Purchase

  • Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, which ensures that constant tension is applied to the media and eliminates the risk of void formation during column use.
  • High-quality media yields excellent lot-to-lot performance.
  • Compatible with SFC upon request.
  • Available in dimensions up to 50mm i.d.

When column performance is past optimal, use the XL repack service:

  • Pay for just the media, not the hardware.
  • Enjoy cost savings over time compared to purchasing standard hardware.
  • Return your column to Thermo Fisher to have it repacked and sent back to you.
  • After completing your purchase, our team will reach out to you with instructions on how to return your column.

Stationary Phase Details

Similar selectivity, less retention than C18 or C8 columns.

  • Outstanding retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column.
  • Best for analysis of hydrophobic polypeptides and proteins.
  • C4 (USP L26) selectivity.

Customer Specific Orders

  • Please contact your local sales representative for Customer Specific Orders.
Specifications
Carbon Load5%
Column FormatPreparative, Xtended Life, Repack
Column TypeReversed Phase
Diameter (Metric)25 mm
EndcappedYes
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
PhaseReversed Phase
Pore Size175 Å
Product LinePrep LC
Stationary PhaseC4
Surface Area (Metric)220 m²/g
Temperature60°C
USP TypeL26
Particle Size5 μm
pH2 to 8
Unit SizeEach