D-(-)-Mannitol, 97.0-102.0% (dry basis), powder, USP, Multi-Compendial, GMP, J.T.Baker™
D-(-)-Mannitol, 97.0-102.0% (dry basis), powder, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

D-(-)-Mannitol, 97.0-102.0% (dry basis), powder, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
2553-01500 g
2553-0712 kg
2553-0950 kg
2553-052,5 kg
Número de catálogo 2553-01
Precio (MXN)
-
Solicite un presupuesto
Cantidad:
500 g
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorBlanco
Punto de fusión164°C to 169°C
EnvasePoly bottle, tamper-evident cap
Punto de ebullición290°C to 295°C (4 hPa)
GradoMulticomponente/USP
Peso molecular (g/mol)182.17
Porcentaje de pureza97 to 102%
Forma físicaPowder
Cantidad500 g
Unit Size500 g