D-(-)-Mannitol, 97.0-102.0% (dry basis), powder, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

D-(-)-Mannitol, 97.0-102.0% (dry basis), powder, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2553-052.5 kg
2553-0712 kg
2553-01500 g
2553-0950 kg
Catalog number 2553-05
Price (EUR)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point164°C to 169°C
PackagingPoly bottle, tamper-evident cap
Boiling Point290°C to 295°C (4 hPa)
GradeMulti-Compendial/USP
Molecular Weight (g/mol)182.17
Percent Purity97 to 102%
Physical FormPowder
Quantity2.5 kg
Unit SizeEach