L(+)-Gulonic acid gamma-lactone, 95+%
L(+)-Gulonic acid gamma-lactone, 95+%
L(+)-Gulonic acid gamma-lactone, 95+%
Thermo Scientific Chemicals

L(+)-Gulonic acid gamma-lactone, 95+%

CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
25557025025 gGlass bottle
Catalog number 255570250
Price (USD)
200.65
Online exclusive
223.00
Save 22.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
200.65
Online exclusive
223.00
Save 22.35
Each
Chemical Identifiers
CAS1128-23-0
IUPAC Name(3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
Molecular FormulaC6H10O6
InChI KeySXZYCXMUPBBULW-SKNVOMKLSA-N
SMILESOC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
HPLC>=95.0 %
Specific optical rotation+51° to +58° (20°C, 589 nm) (c=4, water)
Appearance (Color)White to light beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only