4-Methyl-2-pentanone, 99.5%, for analysis
4-Methyl-2-pentanone, 99.5%, for analysis
4-Methyl-2-pentanone, 99.5%, for analysis
Acros Organics

4-Methyl-2-pentanone, 99.5%, for analysis

CAS: 108-10-1 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Plastic drum
Catalog number 255660100
also known as 25566
HSN 29141300
Price (INR)
53,479.00
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (INR)
53,479.00
Each
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color scale=<15 APHA
GC>=99.4 %
Zinc (Zn)=<0.1 ppm
Residue after evaporation=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only