3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water
3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water
3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water
Thermo Scientific Chemicals

3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water

3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5 | CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 255790250
Price (USD)
51.00
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
51.00
Each
Chemical Identifiers
CAS937-14-4
IUPAC Name3-chlorobenzene-1-carboperoxoic acid
Molecular FormulaC7H5ClO3
InChI KeyNHQDETIJWKXCTC-UHFFFAOYSA-N
SMILESOOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric70 to 75 %
Water% balance (K.F.)
Appearance (Color)White
3-Chlorobenzoic acid=<12 %
HPLCdi-m-Chlorobenzoyl peroxide: < 0.1% Typical
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only