Hypersil GOLD™ Amino HPLC Columns
Thermo Scientific™

Hypersil GOLD™ Amino HPLC Columns

Achieve outstanding peak shape and sensitivity in weak anion exchange, reversed phase and normal phase modes with Hypersil GOLD Amino LC Columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
25702-05213050 mm2.1 mm1.9 μm
25705-05213050 mm2.1 mm5 μm
25705-05463050 mm4.6 mm5 μm
25705-154630150 mm4.6 mm5 μm
25705-254030250 mm4 mm5 μm
25705-254630250 mm4.6 mm5 μm
25702-202130200 mm2.1 mm1.9 μm
25702-152130150 mm2.1 mm1.9 μm
25702-102130100 mm2.1 mm1.9 μm
25703-154630150 mm4.6 mm3 μm
25703-153030150 mm3 mm3 μm
25703-152130150 mm2.1 mm3 μm
25703-03213030 mm2.1 mm3 μm
25703-01400110 mm4 mm3 μm
25703-01300110 mm3 mm3 μm
25705-01400110 mm4 mm5 μm
Catalog number 25702-052130
Price (TWD)
-
Length (Metric):
50 mm
Diameter (Metric):
2.1 mm
Particle Size:
1.9 μm

Achieve excellent chromatographic separations in weak anion exchange, reversed phase and normal phase modes with Thermo Scientific™ Hypersil GOLD™ Amino LC Columns. These columns, featuring a high-performance aminopropyl phase, provide outstanding peak shape and sensitivity and are available with a 1.9μm particle size for high speed, high efficiency separations.

Excellent Chromatographic Properties in Three Modes
  • Retains anions and organic acids in weak anion exchange mode when used with common buffers and an organic modifier
  • Excels for carbohydrate analysis in reversed phase mode
  • Offers an alternative selectivity to silica columns in normal phase chromatography
About the Hypersil GOLD Family of HPLC Columns

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.

  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • Providing 1.9, 3, and 5μm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeMixed Mode
EndcappedYes
Flow Rate0.5 mL/min
For Use WithHILIC
Max. Pressure18130 psi (1250 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseAmino (NH2)
Surface Area220 m²/g
Temperature60°C
USP TypeL8
Carbon Load2%
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Particle Size1.9 μm
PhaseReversed Phase, Normal Phase, Ion Exchange, HILIC
Pore Size175 Å
Product LineHypersil GOLD
TypeAnalytical Column
pH2 to 8
Unit SizeEach