Lauryl chloride, 99%
Lauryl chloride, 99%
Lauryl chloride, 99%
Thermo Scientific Chemicals

Lauryl chloride, 99%

CAS: 112-52-7 | C12H25Cl | 204.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 257102500
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.60
Save 5.95
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.60
Save 5.95
Each
Chemical Identifiers
CAS112-52-7
IUPAC Name1-chlorododecane
Molecular FormulaC12H25Cl
InChI KeyYAYNEUUHHLGGAH-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only