beta-Apo-oxytetracycline, 'can be used as secondary standard'
beta-Apo-oxytetracycline, 'can be used as secondary standard'
beta-Apo-oxytetracycline, 'can be used as secondary standard'
beta-Apo-oxytetracycline, 'can be used as secondary standard'
Thermo Scientific Chemicals

beta-Apo-oxytetracycline, 'can be used as secondary standard'

Beta-apo-oxytetracycline, CAS # 18751-99-0, is a member of the tetracycline family and is a degradation production formed from oxytetracycline. This compound displays antibiotic activity.
Have Questions?
產品號碼QuantityPackaging
257731000100mgGlass bottle
產品號碼 257731000
價格 (TWD)
-
申請報價
Quantity:
100mg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS18751-99-0
IUPAC Name4-{4,5-dihydroxy-9-methyl-3-oxo-1H,3H-naphtho[2,3-c]furan-1-yl}-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohex-1-ene-1-carboxamide
Molecular FormulaC22H22N2O8
InChI KeyDRKMHDAKULCOKQ-UHFFFAOYNA-N
SMILESCN(C)C1C(C2OC(=O)C3=C2C(C)=C2C=CC=C(O)C2=C3O)C(O)C(=O)C(C(N)=O)=C1O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Purity>=80 % (as is, calculated)
Appearance (Color)Yellow-brown to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Beta-apo-oxytetracycline is a degradation product derived from oxytetracycline and displays antibiotic activity
• Similar to its parent compound, oxytetracycline, beta-apo-oxytetracycline can also inhibit protein synthesis by binding to the 30S ribosomal subunit

Applications

• In laboratory assays, it promotes the inhibition of the growth of aerobic sludge bacteria, Pseudomonas, Agrobacterium, Moraxella, Bacillus, as well as tetracycline-resistant strains of E. coli

RUO – Research Use Only

Literature References

Nguyen, V.; Nguyen, V.; Li, C.; Zhou, G. The degradation of oxytetracycline during thermal treatments of chicken and pig meat and the toxic effects of degradation products of oxytetracycline on rats. Journal of Food Science and Technology. 2015, 52, 2842–2850.

Halling-Sørensen, B.; Sengeløv, G.; Tjørnelund, J. Toxicity of tetracyclines and tetracycline degradation products to environmentally relevant bacteria, including selected tetracycline-resistant bacteria. Archives of Environmental Contamination and Toxicology. 2002, 42, 263–271.