Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized
Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized
Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized
Thermo Scientific Chemicals

Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized

Oxidation reagent | CAS: 79-21-0 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 257751000
Price (EUR)
73,65
Online Exclusive
81,20
Save 7,55 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
73,65
Online Exclusive
81,20
Save 7,55 (9%)
Each
Chemical Identifiers
CAS79-21-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow-green
Titration with Na2S2O334.0 to 39.0 % (PAA content)
Appearance (Form)Solution
Titration11.0 to 15.0 % (H2O2 content)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only