Isobutyric anhydride, 97%
Isobutyric anhydride, 97%
Isobutyric anhydride, 97%
Thermo Scientific Chemicals

Isobutyric anhydride, 97%

CAS: 97-72-3 | C8H14O3 | 158.2 g/mol
数量:
500 g
2 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 257780020
価格(JPY)
-
数量:
2 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS97-72-3
IUPAC Name2-methylpropanoyl 2-methylpropanoate
Molecular FormulaC8H14O3
InChI KeyLSACYLWPPQLVSM-UHFFFAOYSA-N
SMILESCC(C)C(=O)OC(=O)C(C)C
さらに表示
Refractive index1.4050 to 1.4070 (20°C, 589 nm)
GC=<1.0 % (Isobutyric acid)
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=96.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only