Tetracosanol, 99+%
Tetracosanol, 99+%
Tetracosanol, 99+%
Thermo Scientific Chemicals

Tetracosanol, 99+%

CAS: 506-51-4 | C24H50O | 354.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 258150010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS506-51-4
IUPAC Nametetracosan-1-ol
Molecular FormulaC24H50O
InChI KeyTYWMIZZBOVGFOV-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Glistening powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only