5-Fluorocytosine, 99+%
5-Fluorocytosine, 99+%
5-Fluorocytosine, 99+%
Thermo Scientific Chemicals

5-Fluorocytosine, 99+%

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 258340010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2022-85-7
IUPAC Name6-amino-5-fluoro-1,2-dihydropyrimidin-2-one
Molecular FormulaC4H4FN3O
InChI KeyXRECTZIEBJDKEO-UHFFFAOYSA-N
SMILESNC1=C(F)C=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Loss on drying=<1.0 % (1 g, 105°C)
Titration with HClO4>=99.0 % (On dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only