(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%
Thermo Scientific Chemicals

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate, 97%

CAS: 37031-30-4 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 258440100
Price (TWD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS37031-30-4
IUPAC Name4,5-dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
Molecular FormulaC9H14O6
InChI KeyROZOUYVVWUTPNG-WDSKDSINSA-N
SMILESCOC(=O)[C@H]1OC(C)(C)O[C@@H]1C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Specific optical rotation+52° to +56° (20°C, 589 nm) (neat)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only