3-Methyl-3-methoxybutanol, 99%
3-Methyl-3-methoxybutanol, 99%
3-Methyl-3-methoxybutanol, 99%
Thermo Scientific Chemicals

3-Methyl-3-methoxybutanol, 99%

CAS: 56539-66-3 | C6H14O2 | 118.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 259752500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56539-66-3
IUPAC Name3-methoxy-3-methylbutan-1-ol
Molecular FormulaC6H14O2
InChI KeyMFKRHJVUCZRDTF-UHFFFAOYSA-N
SMILESCOC(C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only