Isobutyl acetate, 98%
Isobutyl acetate, 98%
Isobutyl acetate, 98%
Thermo Scientific Chemicals

Isobutyl acetate, 98%

CAS: 110-19-0 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 259830010
Price (USD)
60.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.00
Each
Chemical Identifiers
CAS110-19-0
IUPAC Name2-methylpropyl acetate
Molecular FormulaC6H12O2
InChI KeyGJRQTCIYDGXPES-UHFFFAOYSA-N
SMILESCC(C)COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=97.5 %
Density(20°C) 0.871 to 0.875 kg/l
Infrared spectrumConforms
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only