Hypersil GOLD™ Peptide HPLC Column
Thermo Scientific™

Hypersil GOLD™ Peptide HPLC Column

Hypersil GOLD™ Peptide performance is assured by our dual quality confirmation, which is performed on both neutrals compounds as well as on a Cytochrome C digest.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Max. Pressure
26002-152130150 mm21756 psi (1500 bar)
26002-05213050 mm14503 psi (1000 bar)
26002-102130100 mm21756 psi (1500 bar)
Catalog number 26002-152130
Price (HKD)
-
Length (Metric):
150 mm
Max. Pressure:
21756 psi (1500 bar)

Resolve peptide challenges with one column - NEW Hypersil GOLD Peptide.

Achieve consistent retention of all peptides, including hydrophilic ones, and resolve deamidated species. Designed for stability and lot-to-lot consistency, this column meets the needs of biopharmaceutical product development and manufacturing control.

Hypersil GOLD Peptide column performance is assured through dual quality confirmation, including a peptide-based QC test, ensures confidence in ultrapure silica, ligand bonding, and column packing for unparalleled sequence coverage and higher peak capacity compared to other peptide columns.

Consistent, Reliable Results

Enjoy excellent reproducibility with minimal RSD%, ensuring consistency from column-to-column and lot-to-lot.

Optimized Peptide Retention - Retain More, Discover More

Our column uniquely holds onto hydrophilic peptides, ensuring no critical data is missed.

Exceptional Peak Capacity - Separate with Superiority

Achieve higher peak capacity for detailed and clear separations, ensuring every component is distinctly resolved.

Advanced PTM Analysis - Decipher Complex Modifications

Efficiently separate key post-translational modifications like deamidation, crucial for in-depth protein analysis. Excellent peak shape for peptides using either FA- or TFA-containing mobile phases.

Unparalleled Sequence Coverage - See the Full Picture

High sequence coverage provides a comprehensive analysis, revealing the true complexity of your samples.

Specifications
Column FormatAnalytical Column
EndcappedYes
For Use WithUHPLC
For Use With (Application)Peptide
Max. Pressure21756 psi (1500 bar)
Packing MaterialUltrapure Silica
Particle ShapeFully Porous
QuantityEach
Stationary PhaseC18 Selectivity Peptide
Surface Area220 m²/g
Temperature60°C
USP TypeL1
Carbon Load11%
Diameter (Metric)2.1 mm
Length (Metric)150 mm
MaterialStainless Steel
Particle Size1.9 μm
PhaseReversed Phase
Pore Size175 Å
Product LineHypersil GOLD
TechniqueUHPLC, HPLC
pH2 to 11
Unit SizeEach

Frequently asked questions (FAQs)

What application areas or workflows are best served by Hypersil GOLD Peptide columns?

Hypersil GOLD Peptide columns are particularly suited for:
• Biopharmaceutical peptide mapping and manufacturing control (where PTMs, deamidation, and hydrophilic peptides matter).
• LC-MS peptide workflows include digest analysis, proteomics, and glycopeptide variant monitoring.

What mobile phase or buffer considerations should I be aware of when using Hypersil GOLD Peptide columns?

You need to ensure that your sample is fully dissolved and free of particles (e.g., filter 0.2 µm or centrifuge) before injection. The column supports both FA- and TFA-containing mobile phases with excellent peak shape for peptides. For column equilibration, you need to flush new columns with at least 10 column volumes of the new mobile phase. Additionally, take into consideration that at higher pH (>8) and elevated temperature, column lifetime may be shortened.

How do Hypersil GOLD Peptide columns handle hydrophilic peptides, which often elute too early on traditional columns?

Hypersil GOLD Peptide columns are unique in that they hold onto hydrophilic peptides, ensuring no critical data is missed. In other words, it has been engineered (via the packing material, ligand density and pore structure) to provide better retention of less-hydrophobic peptide species compared to generic RP columns, to reduce early elution or loss of critical peptides in a digest map.

What makes Hypersil GOLD Peptide columns specifically suited for peptide analysis?

The Hypersil GOLD Peptide column is built on 1.9 µm, 175 Å pore size spherical, fully porous ultrapure silica, specifically optimized for peptide separations. It provides consistent retention of all peptides—including hydrophilic peptides—and resolves post-translational modifications such as deamidation. Compared with many general-purpose C18 peptide columns, it offers higher peak capacity, better retention of hydrophilic peptides, and enhanced sequence coverage.