Horse Serum, heat inactivated, New Zealand origin
Gibco™

Horse Serum, heat inactivated, New Zealand origin

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
26050070100 mL
26050088500 mL
Catalog number 26050070
Price (USD)
28.65
Online exclusive
29.50
Each
Quantity:
100 mL
Price (USD)
28.65
Online exclusive
29.50
Each

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera Category: Specialty

Origin: New Zealand.

EIA tested (negative) serum from a donor herd.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Product LineGibco
Shipping ConditionFrozen
SpeciesHorse/Equine, Horse
Country of OriginNew Zealand
FormLiquid
Product TypeHorse Serum
Quantity100 mL
Serum TreatmentHeat-inactivated
SterilitySterile
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C, Shipping conditions: Frozen

Frequently asked questions (FAQs)

Does horse serum contain tetracycline?

No. Our horse serum is collected from a controlled herd. Should one of the animals require treatment with an antibiotic, that animal is removed from the drawing program for the full witholding period of any drug administered.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

When should I use heat-inactivated serum?

Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

Citations & References (2)

Citations & References
Abstract
Tyrosine Phosphatase STEP Is a Tonic Brake on Induction of Long-Term Potentiation.
Authors: Pelkey Kenneth A; Askalan Rand; Paul Surojit; Kalia Lorraine V; Nguyen Tri Hung; Pitcher Graham M; Salter Michael W; Lombroso Paul J;
Journal:Neuron
PubMed ID:11931747
'The functional roles of protein tyrosine phosphatases (PTPs) in the developed CNS have been enigmatic. Here we show that striatal enriched tyrosine phosphatase (STEP) is a component of the N-methyl-D-aspartate receptor (NMDAR) complex. Functionally, exogenous STEP depressed NMDAR single-channel activity in excised membrane patches. STEP also depressed NMDAR-mediated synaptic currents ... More
Essential role of cAMP-response element-binding protein activation by A2A adenosine receptors in rescuing the nerve growth factor-induced neurite outgrowth impaired by blockage of the MAPK cascade.
Authors: Cheng Hsiao-Chun; Shih Hsiu-Ming; Chern Yijuang;
Journal:J Biol Chem
PubMed ID:12114502
We found in the present study that stimulation of the A(2A) adenosine receptor (A(2A)-R) using an A(2A)-selective agonist (CGS21680) rescued the blockage of nerve growth factor (NGF)-induced neurite outgrowth when the NGF-evoked MAPK cascade was suppressed by an MEK inhibitor (PD98059) or by a dominant-negative MAPK mutant (dnMAPK). This action ... More