TraceGOLD™ TG-OPP I and II GC Columns
Thermo Scientific™

TraceGOLD™ TG-OPP I and II GC Columns

Analyze organophosphorus pesticides with ultra-low bleed, superior inertness and consistent reproducibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Stationary Phase
26076-14200.25 mmTraceGOLD™ TG-OPP I
26076-22400.32 mmTraceGOLD™ TG-OPP I
26075-14200.25 mmTraceGOLD™ TG-OPP II
26075-57600.32 mmTraceGOLD™ TG-OPP II
Catalog number 26076-1420
Price (BRL)
-
Diameter (Metric):
0.25 mm
Stationary Phase:
TraceGOLD™ TG-OPP I
Analyze organophosphorus pesticides with ultralow bleed, superior inertness, and consistent reproducibility using Thermo Scientific™ TraceGOLD™ TG-OPP I and II GC Columns.This column's low bleed delivers excellent signal-to-noise ratio, sensitivity, and mass spectral integrity. Expect full separation of organophosphorus pesticides with a fast analysis time. TraceGOLD TG-OPP I and II GC Columns are ideal for U.S. EPA methods 8141A.
A Leap Forward in Performance with Low Bleed and Superior Inertness
  • Low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimal column bleed extends column lifetime, leading to higher productivity.
  • Reduced column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Redefines Expectations for GC
  • Low bleed, delivering excellent signal-to-noise ratio, sensitivity and mass spectral integrity.
  • Fast analysis time, facilitating full separation of organophosphorus pesticides.
  • Ideal for U.S. EPA methods 8141A

Similar To
  • Rtx™-OPPesticides
  • Rtx™-OPPesticides2
Specifications
Diameter (Metric)0.25 mm
For Use With (Application)GC/MS for Organophosphorus Pesticides
Max. Temperature (Metric)330°C
PolarityNon-Polar
Diameter (Metric) Inner0.25 mm
Film Thickness0.25 μm
Length (Metric)30 m
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-OPP I
Unit SizeEach