Thermo Scientific™

TraceGOLD TG-POLAR GC Columns

Catalog number: 26082-3360
Have Questions?
Thermo Scientific™

TraceGOLD TG-POLAR GC Columns

Catalog number: 26082-3360
Have Questions?

Refine your expectations for high-polarity GC/MS with the Thermo Scientific™ TraceGOLD™ TG-POLAR columns, which are specifically designed with a polymer and surface treatment to overcome traditional problems with high-polarity columns.

Engineered for Superior Performance

  • Highly polar phase, 90% biscyanopropyl/10% phenylcyanopropyl polysiloxane, not bonded
  • Strong dipole moment and high selectivity for cis/trans compounds or compounds with conjugated double bonds
  • Equivalent to U.S.P G8 and G48
Applications:
  • Cis/Trans fatty acid methyl esters (FAMEs)
  • Dioxins
 
Catalog Number
26082-3360
Unit Size
Each
Diameter (Metric)
0.53 mm
Film Thickness
1.5 μm
Length (Metric)
30 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness1.5 μm
Length (Metric)30 m
PolarityHighly Polar
DescriptionTG-Polar 30 m x 0.53 mm x 1.5 μm
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 μm
Length (Metric)
30 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 μm
Length (Metric)
60 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.1 μm
Length (Metric)
105 m
Price (USD)
Full specifications
Quantity
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)105 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
Catalog Number
Unit Size
Each 1
Diameter (Metric)
0.25 mm
Film Thickness
0.2 μm
Length (Metric)
30 m
Price (USD)
Full specifications
Quantity
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach 1
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.2 μm
Length (Metric)
60 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.25 mm
Film Thickness
0.2 μm
Length (Metric)
105 m
Price (USD)
Full specifications
Quantity
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)105 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.32 mm
Film Thickness
0.2 μm
Length (Metric)
30 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 30 m Length
Unit SizeEach
Catalog Number
Unit Size
Each
Diameter (Metric)
0.32 mm
Film Thickness
0.2 μm
Length (Metric)
60 m
Price (USD)
Full specifications
Quantity
-
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 60 m Length
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDiameter (Metric)Film ThicknessLength (Metric)Price (USD)Quantity
26082-3360Full specifications
Each0.53 mm1.5 μm30 mRequest A Quote-
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness1.5 μm
Length (Metric)30 m
PolarityHighly Polar
DescriptionTG-Polar 30 m x 0.53 mm x 1.5 μm
Unit SizeEach
26082-0470Full specifications
Each0.25 mm0.1 μm30 mRequest A Quote-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach
26082-0590Full specifications
Each0.25 mm0.1 μm60 mRequest A Quote-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
26082-5000Full specifications
Each0.25 mm0.1 μm105 m
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.1 μm
Length (Metric)105 m
PolarityHighly Polar
Description0.1 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
26082-5010Full specifications
Each 10.25 mm0.2 μm30 m
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 30 m Length
Unit SizeEach 1
26082-5020Full specifications
Each0.25 mm0.2 μm60 mRequest A Quote-
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 60 m Length
Unit SizeEach
26082-5030Full specifications
Each0.25 mm0.2 μm105 m
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)105 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 105 m Length
Unit SizeEach
26082-5040Full specifications
Each0.32 mm0.2 μm30 m
-
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)30 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 30 m Length
Unit SizeEach
26082-5050Full specifications
Each0.32 mm0.2 μm60 mRequest A Quote-
Diameter (Metric)0.32 mm
Diameter (Metric) Inner0.32 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Max. Temperature260/275°C
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.32 mm ID; 60 m Length
Unit SizeEach
Showing 9 of 9
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Alternative Uses:
  • Alcohols
  • Aldehydes
  • Biodiesel–FAMEs
Similar To:
  • DB-23
  • HP-23
  • Rtx™™-2330
  • SP™-2330
  • SP™-2380
  • SPB™-2560
  • HP-88
  • Silar™™ 90c
  • CP-Sil 88 FAME
  • CP-Sil 88
  • BPX 70
  • BPX 90
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text