TraceGOLD TG-POLAR GC Columns
Actual product may vary

TraceGOLD TG-POLAR GC Columns

Redefine your expectations for high-polarity GC columns with the Thermo Scientific TraceGold TG-POLAR Columns
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
26082-502060 m0.25 mm0.2 μm
26082-047030 m0.25 mm0.1 μm
26082-059060 m0.25 mm0.1 μm
26082-5000105 m0.25 mm0.1 μm
26082-501030 m0.25 mm0.2 μm
26082-5030105 m0.25 mm0.2 μm
26082-504030 m0.32 mm0.2 μm
26082-505060 m0.32 mm0.2 μm
Catalog number 26082-5020
HSN 84212900
Price (INR)
153,109.00
Each
Length (Metric):
60 m
Diameter (Metric) Inner:
0.25 mm
Film Thickness:
0.2 μm
Price (INR)
153,109.00
Each

Refine your expectations for high-polarity GC/MS with the Thermo Scientific™ TraceGOLD™ TG-POLAR columns, which are specifically designed with a polymer and surface treatment to overcome traditional problems with high-polarity columns.

Engineered for Superior Performance

  • Highly polar phase, 90% biscyanopropyl/10% phenylcyanopropyl polysiloxane, not bonded
  • Strong dipole moment and high selectivity for cis/trans compounds or compounds with conjugated double bonds
  • Equivalent to U.S.P G8 and G48

Applications:

  • Cis/Trans fatty acid methyl esters (FAMEs)
  • Dioxins
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Alternative Uses:
  • Alcohols
  • Aldehydes
  • Biodiesel–FAMEs
Similar To:
  • DB-23
  • HP-23
  • Rtx™™-2330
  • SP™-2330
  • SP™-2380
  • SPB™-2560
  • HP-88
  • Silar™™ 90c
  • CP-Sil 88 FAME
  • CP-Sil 88
  • BPX 70
  • BPX 90
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Film Thickness0.2 μm
Length (Metric)60 m
PolarityHighly Polar
Description0.2 um Thickness; 0.25 mm ID; 60 m Length
Diameter (Metric)0.25 mm
USP TypeG8, G48
Stationary PhaseTraceGOLD™ TG-Polar
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Max. Temperature (Metric)260/275°C
Unit SizeEach