Hypersil GOLD™ AX HPLC Columns
Actual product may vary
Hypersil GOLD™ AX HPLC Columns
Hypersil GOLD™ AX HPLC Columns
Thermo Scientific™

Hypersil GOLD™ AX HPLC Columns

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
26103-151030150 mm1 mm3 μm
26103-03213030 mm2.1 mm3 μm
26103-102130100 mm2.1 mm3 μm
26103-152130150 mm2.1 mm3 μm
26103-01400110 mm4 mm3 μm
26103-01210110 mm2.1 mm3 μm
Catalog number 26103-151030
Price (USD)
-
Length (Metric):
150 mm
Diameter (Metric):
1 mm
Particle Size:
3 μm

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.

Excellent Chromatographic Performance
  • Outstanding peak shape and selectivity
  • Superb stability under demanding pH conditions
  • Higher efficiency than polymer-based ion exchange columns
  • Improved speed with 1.9μm particle size columns
About the Hypersil GOLD Family of HPLC Columns

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeMixed Mode
EndcappedNo
Flow Rate0.05 mL/min
For Use WithHILIC
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhasePolymeric Amine
Surface Area220 m²/g
Temperature60°C
Carbon Load6%
Diameter (Metric)1 mm
Length (Metric)150 mm
Particle Size3 μm
PhaseHILIC and Ion Exchange
Pore Size175 Å
Product LineHypersil GOLD
TypeAnalytical Column
pH2 to 8
Unit SizeEach