Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Diameter (Metric) | Particle Size |
---|---|---|---|
26105-252130 | 250 mm | 2.1 mm | 5 μm |
26103-102130 | 100 mm | 2.1 mm | 3 μm |
26103-012101 | 10 mm | 2.1 mm | 3 μm |
Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.