Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™
Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™
Avantor™

Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets FCC, ACS and NF specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 lb.
47 lb.
500 lb.
Packaging:
View all
Poly bottle, pressure bottom
Catalog number 2612-06
Price (MXN)
-
Request A Quote
Quantity:
10 lb.
Packaging:
Poly bottle, pressure bottom
This product is GMP Manufactured and meets FCC, ACS and NF specifications.
This product is GMP Manufactured and meets FCC, ACS and NF specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point110°C (1013 hPa)
CAS7647-01-0
Chemical Name or MaterialHydrochloric acid
ColorColorless
Melting Point-30°C
Molecular FormulaHCl
PackagingPoly bottle, pressure bottom
SynonymHydrochloric acid, Hydrogen chloride solution
GradeNF, FCC, ACS
Molecular Weight (g/mol)36.46
Percent Purity36.5% to 38%
Quantity10 lb.
Unit SizeEach