Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™
Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™
Avantor™

Hydrochloric acid, 36.5-38.0%, NF, FCC, ACS, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets FCC, ACS and NF specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
2.5 L
10 lb.
47 lb.
500 lb.
Envase:
Ver todo
Polyethylene drum
Número de catálogo 2612-19
Precio (USD)
-
Solicite un presupuesto
Cantidad:
500 lb.
Envase:
Polyethylene drum
This product is GMP Manufactured and meets FCC, ACS and NF specifications.
This product is GMP Manufactured and meets FCC, ACS and NF specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición110°C (1013 hPa)
CAS7647-01-0
Nombre del producto químico o materialHydrochloric acid
ColorColorless
Punto de fusión-30°C
Fórmula molecularHCl
EnvasePolyethylene drum
SinónimoHydrochloric acid, Hydrogen chloride solution
GradoNF, FCC, ACS
Peso molecular (g/mol)36.46
Porcentaje de pureza36.5% to 38%
Cantidad500 lb.
Unit SizeEach