Search
View additional product information for MT(PEG)4 Methyl-PEG-Thiol Compound - FAQs (26132)
2 product FAQs found
MT(PEG)4 Methyl-PEG-Thiol Compound (Cat. No. 26132) turns to liquid at RT. PEG-thiol reagents are low-melting solids that are difficult to weigh and dispense. To facilitate handling, make a stock solution by dissolving the entire contents of the bottle (100 mg in solid or liquid form) in dimethylsulfoxide (DMS) or dimethylformamide (DMF) to the desired stock concentration. Store unused stock solution at -20 degrees C.
Please note: the molecular weight of MT(PEG)4 Methyl-PEG-Thiol Compound is 224.32 g/ml.
For more information about protocol an use, please refer to the Instructions.
MT(PEG)4 Methyl-PEG-Thiol Compound (Cat. No. 26132) turns to liquid at RT. It is normal for the product to change state during transport as it is shipped at ambient temperature. The PEG-thiol reagents are low-melting solids that are difficult to weigh and dispense. To facilitate handling, make a stock solution by dissolving the entire contents of the bottle (100 mg in solid or liquid form) in dimethylsulfoxide (DMS) or dimethylformamide (DMF) to the desired stock concentration. Store unused stock solution at -20 degrees C.
Please note: the molecular weight of MT(PEG)4 Methyl-PEG-Thiol Compound is 224.32 g/ml.
For more information about protocol and use, please refer to the Instructions.