Order this product, then join the Aspire Member Program to receive 3X points on this product. This is a limited time offer in USA & Canada only.
Gibco™

Fetal Bovine Serum, qualified, One Shot™ format, US origin This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  A3160502

 Related applications:

Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Sera category: Performance
Origin: United States.
Endotoxin level: ≤10 EU/mL
Hemoglobin level: ≤25 mg/dL

Aliquot-free 50 mL bottle
The Gibco™ One Shot™ 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:
• Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
• Engineered to freeze/thaw without splitting or breaking
• Fits in test tube racks and frees up freezer space
• No labeling mishaps with our easy-to-read package label containing all of the information you need
• 33% less waste generated than aliquoting from 500 mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

Specifications

Serum Treatment: Standard (Sterile-Filtered)
Purity or Quality Grade: Qualified
Country of Origin: US
Format: One Shot
Product Line: One Shot™
Product Size: 10 x 50 mL
Green Features: Fewer resources used and less waste
Regulatory Statement: For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols