SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate) - FAQs
SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate) - FAQs
View additional product information for SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate) - FAQs (26167)
1 product FAQs found
Can you provide the shelf-life for SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate)?
SDA (NHS-Diazirine) (succinimidyl 4,4'-azipentanoate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.