Bovine Serum, heat inactivated, New Zealand origin, 100 mL - FAQs
View additional product information for Bovine Serum, heat inactivated, New Zealand origin - FAQs (26170035, 26170043)
2 product FAQs found
When should I use heat-inactivated serum?
Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.