Cyclopropylamine, 98%
Cyclopropylamine, 98%
Cyclopropylamine, 98%
Thermo Scientific Chemicals

Cyclopropylamine, 98%

CAS: 765-30-0 | C3H7N | 57.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 263681000
Price (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS765-30-0
IUPAC Namecyclopropanamine
Molecular FormulaC3H7N
InChI KeyHTJDQJBWANPRPF-UHFFFAOYSA-N
SMILESNC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only