3-Chloro-1H-indazole, 99%
3-Chloro-1H-indazole, 99%
3-Chloro-1H-indazole, 99%
Thermo Scientific Chemicals

3-Chloro-1H-indazole, 99%

CAS: 29110-74-5 | C7H5ClN2 | 152.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 263740250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS29110-74-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Powder
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only