Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution
Thermo Scientific Chemicals

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 263820010
Price (EUR)
152,00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
152,00
Each
Chemical Identifiers
CAS8001-54-5
Molecular FormulaC9H13ClN
SMILES[Cl-].C[N+](C)(*)CC1=CC=CC=C1
Molecular Weight (g/mol)170.66
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
pH6 to 9 (10 % in water)
Titration Argentometric49 to 52 % (based on M.W.= 352.5)
Impurity=<2 % Free amine + amine HCl (M.W.= 226/262.5)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only