Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Thermo Scientific Chemicals

Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)

CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
檢視全部
Plastic drum
產品號碼 263830100
價格 (TWD)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
化學識別
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
檢視更多
規格Specification Sheet規格表
Particle size=<1 % Residue on 150 µm (>100 mesh) sieve

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only