ES Series Combination Lab Refrigerator/Freezer, 230V - FAQs
View additional product information for ES Series Combination Lab Refrigerator/Freezer, 230V - FAQs (263C-AEV-TS)
2 product FAQs found
Can I change the freezer setpoint for my ES Series Combination Lab Refrigerator/Freezer, 230V (Cat. No. 263C-AEV-TS)?
The freezer setpoint cannot be set. The ES Series Combination Lab Refrigerator/Freezer uses the same compressor as the refrigerator.
If I order an ES Series Lab Refrigerator or Freezer, what type of plug will it come with?
The ES Series Lab Refrigerator or Freezer will be delivered with the compatible plug type and adapter based on where the unit is shipped.