Search
View additional product information for ES Series FMS Combination Refrigerator/Freezers - FAQs (263C-AXV-TS)
1 product FAQs found
The ES Series Lab Refrigerator or Freezer will be delivered with the compatible plug type and adapter based on where the unit is shipped.