Nunc™ Microplate Lids, 96 plates, sterile - FAQs
View additional product information for Nunc™ Microplate Lids - FAQs (253623, 264122, 264612, 264623, 250003, 264611, 250002, 264616, 263339, 250005, 253624)
1 product FAQs found
Which Nunc products can I use for high-throughput, plate-based DNA plasmid minipreps?
We recommend using the following Nunc products:
- 96-Well Polypropylene DeepWell Storage Plate, 2 mL (Cat. No. 278743) (sterile growth plate)
- Microplate Lid for 96 microwell plate (Cat. No. 264122) (to cover plate during growth)
- Polyolefin Acrylate Sealing Tape (Cat. No. 232701) (to cover plate during mixing)
- Fritted 96-Well Deep Well Filter Plate (Cat. No. 278011) (for lysate clarification)
- Fritted 96-Well Deep Well Filter Plate with Glass Fiber Filter (Cat. No. 278010) (for DNA binding)
- 96-Well Polypropylene DeepWell Storage Plate, 1.3 mL (Cat. No. 260252) (reception plate)