1-Chloroadamantane, 98%
1-Chloroadamantane, 98%
1-Chloroadamantane, 98%
Thermo Scientific Chemicals

1-Chloroadamantane, 98%

CAS: 935-56-8 | C10H15Cl | 170.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 264242500
Price (USD)
1,127.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,127.00
Each
Chemical Identifiers
CAS935-56-8
IUPAC Name1-chloroadamantane
Molecular FormulaC10H15Cl
InChI KeyOZNXTQSXSHODFR-UHFFFAOYSA-N
SMILESClC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light grey
Appearance (Form)Adhering crystalline solid and/or chunks
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only