Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Thermo Scientific™

Nunc™ Storage Vials with Caps

Green features
Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) Working
2642620.5 mL
2643002.0 mL
Catalog number 264262
Price (USD)
347.40
Online exclusive
405.00
Save 57.60
Case of 500
Volume (Metric) Working:
0.5 mL
Price (USD)
347.40
Online exclusive
405.00
Save 57.60
Case of 500

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

Specifications
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Closure MaterialPolyethylene
For Use With (Equipment)264263, 270972 Rack
Green FeaturesACT Ecolabel (Gold)
No. per Case500
No. per Pack500
Product TypeStorage Vial with Cap
Volume (Metric) Total0.9 mL
Volume (Metric) Working0.5 mL
Closure TypeScrew Cap
MaterialPPCO
Product LineNunc
Quantity500 Cs.
SterilitySterile
Volume (Metric)0.9 mL
Unit SizeCase of 500